accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 656-65-5

IUPAC Name: 4-bromo-3-fluoroaniline
Synonyms: 4-bromo-3-fluoroaniline
Molecular Weight (g/mol): 190.02
Molecular Formula: C6H5BrFN
InChi Key: YTMVYYAKOPIJCZ-UHFFFAOYSA-N
SMILES: NC1=CC=C(Br)C(F)=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)