accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 656-66-6

IUPAC Name: 3-fluoro-4-iodoaniline
Synonyms: 3-fluoro-4-iodoaniline
Molecular Weight (g/mol): 237.02
Molecular Formula: C6H5FIN
InChi Key: KUVVJHBHRIXJKI-UHFFFAOYSA-N
SMILES: NC1=CC(F)=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results