CAS RN 6573-11-1
IUPAC Name:
1,4,7-trithionane
Synonyms:
1,4,7-trithiacyclononane
Molecular Weight (g/mol):
180.34
Molecular Formula:
C6H12S3
InChi Key:
PQNPKQVPJAHPSB-UHFFFAOYSA-N
SMILES:
C1CSCCSCCS1
1
–
1
of
1
results
1
1,4,7-Trithiacyclononane 98.0+%, TCI America™
CAS: 6573-11-1 Molecular Formula: C6H12S3 Molecular Weight (g/mol): 180.34 InChI Key: PQNPKQVPJAHPSB-UHFFFAOYSA-N PubChem CID: 373835 ChEBI: CHEBI:37418 IUPAC Name: 1,4,7-trithionane SMILES: C1CSCCSCCS1