accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65739-04-0

IUPAC Name: 2-fluoro-N,N-dimethyl-4-nitroaniline
Synonyms: 2-fluoro-N,N-dimethyl-4-nitroaniline
Molecular Weight (g/mol): 184.17
Molecular Formula: C8H9FN2O2
InChi Key: QTXUWCWOGLFTTP-UHFFFAOYSA-N
SMILES: CN(C)C1=C(F)C=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results