accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65799-47-5

IUPAC Name: dimethoxy(methyl){3-[(oxiran-2-yl)methoxy]propyl}silane
Synonyms: dimethoxy(methyl)[3-(oxiran-2-ylmethoxy)propyl]silane
Molecular Weight (g/mol): 220.34
Molecular Formula: C9H20O4Si
InChi Key: WHGNXNCOTZPEEK-UHFFFAOYNA-N
SMILES: CO[Si](C)(CCCOCC1CO1)OC
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)