accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSF

CAS RN 658-28-6

IUPAC Name: 1-fluoro-3-(methylsulfanyl)benzene
Synonyms: 1-fluoro-3-(methylsulfanyl)benzene
Molecular Weight (g/mol): 142.19
Molecular Formula: C7H7FS
InChi Key: SQXSNYMCORGWCE-UHFFFAOYSA-N
SMILES: CSC1=CC=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results