accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 658-79-7

IUPAC Name: (2S)-2-(2-aminoacetamido)-3-(4-hydroxyphenyl)propanoic acid
Synonyms: glycyl tyrosine
Molecular Weight (g/mol): 238.24
Molecular Formula: C11H14N2O4
InChi Key: XBGGUPMXALFZOT-IMWMWJONNA-N
SMILES: NCC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)