accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2ONH2ONHSHN

CAS RN 65802-56-4

IUPAC Name: 6-amino-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one hydrate
Synonyms: 6-amino-2-sulfanylidene-1,3-dihydropyrimidin-4-one hydrate
Molecular Weight (g/mol): 161.18
Molecular Formula: C4H7N3O2S
InChi Key: BWYGMIYEADAIGW-UHFFFAOYSA-N
SMILES: O.NC1=CC(=O)NC(=S)N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results