accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHONNCH3CH3SOONCH3ClHNO(E/Z)

CAS RN 658084-23-2

IUPAC Name: N-(3-chlorophenyl)-3-{[3,5-dimethyl-4-(4-methylpiperazine-1-carbonyl)-1H-pyrrol-2-yl]methylidene}-N-methyl-2-oxo-2,3-dihydro-1H-indole-5-sulfonamide
Synonyms: N-(3-chlorophenyl)-3-{[3,5-dimethyl-4-(4-methylpiperazine-1-carbonyl)-1H-pyrrol-2-yl]methylidene}-N-methyl-2-oxo-1H-indole-5-sulfonamide
Molecular Weight (g/mol): 568.09
Molecular Formula: C28H30ClN5O4S
InChi Key: FPYJSJDOHRDAMT-UHFFFAOYSA-N
SMILES: CN(C1=CC(Cl)=CC=C1)S(=O)(=O)C1=CC2=C(NC(=O)C2=CC2=C(C)C(C(=O)N3CCN(C)CC3)=C(C)N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results