accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65826-95-1

IUPAC Name: 5-methyl-2,3-dihydro-1H-indole
Synonyms: 5-methyl-2,3-dihydro-1H-indole
Molecular Weight (g/mol): 133.19
Molecular Formula: C9H11N
InChi Key: JFUAVVHABJWSFX-UHFFFAOYSA-N
SMILES: CC1=CC2=C(NCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results