accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65854-91-3

IUPAC Name: N-(4-chlorophenyl)-2,2-dimethylpropanamide
Synonyms: N-(4-chlorophenyl)-2,2-dimethylpropanamide
Molecular Weight (g/mol): 211.69
Molecular Formula: C11H14ClNO
InChi Key: IZISMXMXCLUHGI-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)NC1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results