accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClZnH3CHNSHNCH3CH3NH3C

CAS RN 6586-05-6

IUPAC Name: zinc(2+) 3,7-bis(ethylamino)-2,8-dimethyl-5λ⁴-phenothiazin-5-ylium trichloride
Synonyms: new methylene blue
Molecular Weight (g/mol): 484.19
Molecular Formula: C18H22Cl3N3SZn
InChi Key: GGKWUSWRGRCLEV-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Zn++].CCNC1=CC2=[S+]C3=CC(NCC)=C(C)C=C3N=C2C=C1C
Color 
  • (1)
  • (1)
  • (4)
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)