accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3FF

CAS RN 65864-64-4

IUPAC Name: (difluoromethyl)trimethylsilane
Synonyms: (difluoromethyl)trimethylsilane
Molecular Weight (g/mol): 124.21
Molecular Formula: C4H10F2Si
InChi Key: OOKFLLNDYNWCHK-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)