accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65872-41-5

CH3ONOOSH2NN(Z)

CAS RN 65872-41-5

IUPAC Name: (2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetate
Synonyms: (2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetate
Molecular Weight (g/mol): 200.19
Molecular Formula: C6H6N3O3S
InChi Key: NLARCUDOUOQRPB-WTKPLQERSA-M
SMILES: CO\N=C(/C([O-])=O)C1=CSC(N)=N1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)