accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65874-27-3

CH3H3CCH3OOO

CAS RN 65874-27-3

IUPAC Name: tert-butyl 4-formylbenzoate
Synonyms: tert-butyl 4-formylbenzoate
Molecular Weight (g/mol): 206.24
Molecular Formula: C12H14O3
InChi Key: DUNFNBQQWYQKFE-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)C1=CC=C(C=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results