accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CSCH3NH3C

CAS RN 65894-83-9

IUPAC Name: 4,5-dimethyl-2-(2-methylpropyl)-2,5-dihydro-1,3-thiazole
Synonyms: 4,5-dimethyl-2-(2-methylpropyl)-2,5-dihydro-1,3-thiazole
Molecular Weight (g/mol): 171.3
Molecular Formula: C9H17NS
InChi Key: FDOISHJOXPONIV-UHFFFAOYNA-N
SMILES: CC(C)CC1SC(C)C(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results