accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65896-11-9

IUPAC Name: 2-bromo-6-fluoroaniline
Synonyms: 2-bromo-6-fluoroaniline
Molecular Weight (g/mol): 190.02
Molecular Formula: C6H5BrFN
InChi Key: ALZFPYUPNVLVQM-UHFFFAOYSA-N
SMILES: NC1=C(F)C=CC=C1Br
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results