accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65896-16-4

IUPAC Name: N-(2-bromo-6-fluorophenyl)-4,5-dihydro-1H-imidazol-2-amine
Synonyms: romifidine 罗米非定
Molecular Weight (g/mol): 258.09
Molecular Formula: C9H9BrFN3
InChi Key: KDPNLRQZHDJRFU-UHFFFAOYSA-N
SMILES: FC1=CC=CC(Br)=C1NC1=NCCN1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results