accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOFFFOFFFOFFF

CAS RN 659-18-7

IUPAC Name: tris(2,2,2-trifluoroethyl) borate
Synonyms: tris(2,2,2-trifluoroethyl) borate
Molecular Weight (g/mol): 307.91
Molecular Formula: C6H6BF9O3
InChi Key: DIEXQJFSUBBIRP-UHFFFAOYSA-N
SMILES: FC(F)(F)COB(OCC(F)(F)F)OCC(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (1)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results