accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOCH3CH3

CAS RN 659-70-1

IUPAC Name: 3-methylbutyl 3-methylbutanoate
Synonyms: apple oil isoamyl isovalerianate isovalerate isopentyl alcohol isopentyl alcohol, isovalerate isoamyl isovalerate solusterol isoamyl isovalerate (natural) apple essence
Molecular Weight (g/mol): 172.27
Molecular Formula: C10H20O2
InChi Key: XINCECQTMHSORG-UHFFFAOYSA-N
SMILES: CC(C)CCOC(=O)CC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results