accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6590-95-0

IUPAC Name: 1,3-dichloro-2-isothiocyanatobenzene
Synonyms: 1,3-dichloro-2-isothiocyanatobenzene
Molecular Weight (g/mol): 204.07
Molecular Formula: C7H3Cl2NS
InChi Key: SUCGVQHNGIQXGD-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Cl)=C1N=C=S
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)