accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CH3CCH3OONHNH2

CAS RN 65915-94-8

IUPAC Name: hydrogen tert-butyl N-(6-aminohexyl)carbamate chloride
Synonyms: hydrogen tert-butyl N-(6-aminohexyl)carbamate chloride
Molecular Weight (g/mol): 252.78
Molecular Formula: C11H25ClN2O2
InChi Key: JSBWQIZQJOQPFN-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(C)(C)OC(=O)NCCCCCCN
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)