accessibility menu, dialog, popup

UserName

Viewing profile as user:
A(a)

CAS RN 65945-33-7

IUPAC Name: 1,2-diethenylbenzene; bis(2-aminoethyl)[(2-ethenylphenyl)methyl]amine
Synonyms: bis(2-aminoethyl)[(2-ethenylphenyl)methyl]amine; divinylbenzene
Molecular Weight (g/mol): 349.52
Molecular Formula: C23H31N3
InChi Key: UUZFYSHIFAAYRA-UHFFFAOYSA-N
SMILES: C=CC1=CC=CC=C1C=C.NCCN(CCN)CC1=CC=CC=C1C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)