accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 6596-40-3

IUPAC Name: pentacontane
Synonyms: pentacontane
Molecular Weight (g/mol): 703.37
Molecular Formula: C50H102
InChi Key: PFLUOWJPZLHUEA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (4)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)