CAS RN 65973-52-6
IUPAC Name:
methyl 4,6-dichloropyridine-3-carboxylate
Synonyms:
methyl 4,6-dichloropyridine-3-carboxylate
Molecular Weight (g/mol):
206.02
Molecular Formula:
C7H5Cl2NO2
InChi Key:
JEJMDUMJSZTJTI-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CN=C(Cl)C=C1Cl
1
–
1
of
1
results
1
Promotions Available
Methyl 4,6-dichloronicotinate, 95%
Methyl 4,6-dichloronicotinate, 95%
CAS: 65973-52-6 Molecular Formula: C7H5Cl2NO2 Molecular Weight (g/mol): 206.02 MDL Number: MFCD04125732 InChI Key: JEJMDUMJSZTJTI-UHFFFAOYSA-N PubChem CID: 2764083 IUPAC Name: methyl 4,6-dichloropyridine-3-carboxylate SMILES: COC(=O)C1=CN=C(Cl)C=C1Cl