accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 659742-21-9

IUPAC Name: (6-methylpyridin-3-yl)boronic acid
Synonyms: 6-methylpyridin-3-ylboronic acid
Molecular Weight (g/mol): 136.95
Molecular Formula: C6H8BNO2
InChi Key: MZUSCPDSQJSBSY-UHFFFAOYSA-N
SMILES: CC1=NC=C(C=C1)B(O)O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)