CAS RN 65995-63-3
CAS RN 65995-63-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Punicalagin, ≥85% (HPLC), MilliporeSigma™ Supelco™
Punicalagin is a water soluble ellagitannin found in pomegranate juice with high antioxidant activity.