accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOOOHN(S)(S)(R)(R)N

CAS RN 66-32-0

IUPAC Name: (1S,18S,20R,21R)-9-oxo-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2,4,6,14-tetraen-17-ium nitrate
Synonyms: (1S,18S,20R,21R)-9-oxo-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2,4,6,14-tetraen-17-ium nitrate
Molecular Weight (g/mol): 397.43
Molecular Formula: C21H23N3O5
InChi Key: JWKVPTXNWFCSIW-OFWMIIDDSA-O
SMILES: [O-][N+]([O-])=O.O=C1CC2OCC=C3C[NH+]4CC[C@@]56[C@@H]4C[C@@H]3[C@@H]2C5N1C1=CC=CC=C61
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results