accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 660-88-8

IUPAC Name: 5-aminopentanoic acid
Synonyms: 5-aminovaleric acid delta-aminovaleric acid delta-amino-n-valeric acid delta-amino-n-valerate delta-aminovalerate
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: JJMDCOVWQOJGCB-UHFFFAOYSA-N
SMILES: NCCCCC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)