accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66036-77-9

OHOOOH2NNHNNH2N(R)(E)

CAS RN 66036-77-9

IUPAC Name: (2R)-2-amino-5-[(E)-[amino(nitroamino)methylidene]amino]pentanoic acid
Synonyms: (2R)-2-amino-5-[(E)-[amino(nitroamino)methylidene]amino]pentanoic acid
Molecular Weight (g/mol): 219.2
Molecular Formula: C6H13N5O4
InChi Key: MRAUNPAHJZDYCK-SCSAIBSYSA-N
SMILES: N[C@H](CCC\N=C(/N)N[N+]([O-])=O)C(O)=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)