accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6609-56-9

IUPAC Name: 2-methoxybenzonitrile
Synonyms: 2-methoxy-benzonitrile
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: FSTPMFASNVISBU-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C#N
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (7)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)