CAS RN 661-82-5
IUPAC Name:
3-chloro-2,2,3,3-tetrafluoropropanoic acid
Synonyms:
3-chloro-2,2,3,3-tetrafluoropropanoic acid
Molecular Weight (g/mol):
180.48
Molecular Formula:
C3HClF4O2
InChi Key:
XJRMCFMZGRJXEZ-UHFFFAOYSA-N
SMILES:
OC(=O)C(F)(F)C(F)(F)Cl
1
–
1
of
1
results
1
3-Chloro-2,2,3,3-tetrafluoropropionic Acid 98.0+%, TCI America™
CAS: 661-82-5 Molecular Formula: C3HClF4O2 Molecular Weight (g/mol): 180.483 MDL Number: MFCD00153099 InChI Key: XJRMCFMZGRJXEZ-UHFFFAOYSA-N PubChem CID: 2782491 IUPAC Name: 3-chloro-2,2,3,3-tetrafluoropropanoic acid SMILES: C(=O)(C(C(F)(F)Cl)(F)F)O