accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66121-69-5

IUPAC Name: 1-methyl-1H-imidazole-4-carbonitrile
Synonyms: 1-methylimidazole-4-carbonitrile
Molecular Weight (g/mol): 107.12
Molecular Formula: C5H5N3
InChi Key: LAKVILSENFFPRR-UHFFFAOYSA-N
SMILES: CN1C=NC(=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results