accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrN

CAS RN 66127-01-3

IUPAC Name: 3-bromo-1,10-phenanthroline
Synonyms: 3-bromo-1,10-phenanthroline
Molecular Weight (g/mol): 259.11
Molecular Formula: C12H7BrN2
InChi Key: OADZHFGJIVKDJN-UHFFFAOYSA-N
SMILES: BrC1=CN=C2C(C=CC3=CC=CN=C23)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results