accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3ClO

CAS RN 6613-44-1

IUPAC Name: 3,5-dimethylbenzoyl chloride
Synonyms: 3,5-dimethylbenzoyl chloride
Molecular Weight (g/mol): 168.62
Molecular Formula: C9H9ClO
InChi Key: ZJIOBDJEKDUUCI-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1)C(Cl)=O
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)