accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COPHOOOHH3CH3CCH3CH3H3CCH3

CAS RN 66165-37-5

IUPAC Name: [(3,5-di-tert-butyl-4-hydroxyphenyl)methyl](ethoxy)phosphinic acid
Synonyms: (3,5-di-tert-butyl-4-hydroxyphenyl)methyl(ethoxy)phosphinic acid
Molecular Weight (g/mol): 328.39
Molecular Formula: C17H29O4P
InChi Key: WBSRIXCTCFFHEF-UHFFFAOYNA-N
SMILES: CCOP(O)(=O)CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results