accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66171-50-4

CH3OONHO

CAS RN 66171-50-4

IUPAC Name: methyl 6-oxo-1,6-dihydropyridine-3-carboxylate
Synonyms: methyl 6-oxo-1H-pyridine-3-carboxylate
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: JTVVPKLHKMKWNN-UHFFFAOYSA-N
SMILES: COC(=O)C1=CNC(=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results