accessibility menu, dialog, popup

UserName

Viewing profile as user:
OFFFFFFOHHO

CAS RN 662-22-6

IUPAC Name: 3,3,3-trifluoro-2-hydroxy-2-(trifluoromethyl)propanoic acid
Synonyms: 3,3,3-trifluoro-2-hydroxy-2-(trifluoromethyl)propanoic acid
Molecular Weight (g/mol): 212.05
Molecular Formula: C4H2F6O3
InChi Key: CMQUGOHGJUTDGZ-UHFFFAOYSA-N
SMILES: OC(=O)C(O)(C(F)(F)F)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)