accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66230-04-4

IUPAC Name: (R)-cyano(3-phenoxyphenyl)methyl (2R)-2-(4-chlorophenyl)-3-methylbutanoate
Synonyms: esfenvalerate
Molecular Weight (g/mol): 419.91
Molecular Formula: C25H22ClNO3
InChi Key: NYPJDWWKZLNGGM-BJKOFHAPSA-N
SMILES: CC(C)[C@@H](C(=O)O[C@@H](C#N)C1=CC=CC(OC2=CC=CC=C2)=C1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results