CAS RN 66246-88-6
IUPAC Name:
1-[2-(2,4-dichlorophenyl)pentyl]-1H-1,2,4-triazole
Synonyms:
penconazole
Molecular Weight (g/mol):
284.18
Molecular Formula:
C13H15Cl2N3
InChi Key:
WKBPZYKAUNRMKP-UHFFFAOYNA-N
SMILES:
CCCC(CN1C=NC=N1)C1=CC=C(Cl)C=C1Cl
1
–
1
of
1
results
1
TraceCERT™ Penconazol, Certified Reference Material, MilliporeSigma™ Supelco™
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI. Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.