accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClNOO

CAS RN 6627-53-8

IUPAC Name: 4-chloro-2-methoxy-1-nitrobenzene
Synonyms: 4-chloro-2-methoxy-1-nitrobenzene
Molecular Weight (g/mol): 187.58
Molecular Formula: C7H6ClNO3
InChi Key: ABEUJUYEUCCZQF-UHFFFAOYSA-N
SMILES: COC1=CC(Cl)=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results