accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6627-88-9

IUPAC Name: 2,6-dimethoxy-4-(prop-2-en-1-yl)phenol
Synonyms: 2,6-dimethoxy-4-(prop-2-en-1-yl)phenol
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: FWMPKHMKIJDEMJ-UHFFFAOYSA-N
SMILES: COC1=CC(CC=C)=CC(OC)=C1O
Boiling Point 
  • (4)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)