accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66270-97-1

IUPAC Name: 2-oxo-2-phenylethyl 2-[4-(bromomethyl)phenyl]acetate
Synonyms: 2-oxo-2-phenylethyl 2-[4-(bromomethyl)phenyl]acetate
Molecular Weight (g/mol): 347.21
Molecular Formula: C17H15BrO3
InChi Key: BUQPIKAEKYNDAS-UHFFFAOYSA-N
SMILES: BrCC1=CC=C(CC(=O)OCC(=O)C2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results