accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONH2N

CAS RN 6628-77-9

IUPAC Name: 6-methoxypyridin-3-amine
Synonyms: 5-amino-2-methoxypyridine
Molecular Weight (g/mol): 124.14
Molecular Formula: C6H8N2O
InChi Key: UUVDJIWRSIJEBS-UHFFFAOYSA-N
SMILES: COC1=CC=C(N)C=N1
Boiling Point 
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (3)
  • (3)
  • (4)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (5)
  • (3)