accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6628-86-0

IUPAC Name: 5-chloro-2-nitrobenzaldehyde
Synonyms: 5-chloro-2-nitrobenzaldehyde
Molecular Weight (g/mol): 185.56
Molecular Formula: C7H4ClNO3
InChi Key: SWGPIDCNYAYXMJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Cl)C=C1C=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results