accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66309-69-1

IUPAC Name: dihydrogen (6R,7R)-7-[2-(2-amino-1,3-thiazol-4-yl)acetamido]-3-[({1-[2-(dimethylamino)ethyl]-1H-1,2,3,4-tetrazol-5-yl}sulfanyl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid dichloride
Synonyms: dihydrogen cefotiam dichloride
Molecular Weight (g/mol): 598.54
Molecular Formula: C18H25Cl2N9O4S3
InChi Key: BWRRTAXZCKVRON-DGPOFWGLSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CN(C)CCN1N=NN=C1SCC1=C(N2[C@H](SC1)[C@H](NC(=O)CC1=CSC(N)=N1)C2=O)C(O)=O
Color 
  • (2)
Quantity 
  • (1)
  • (1)
Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results