accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOSNH2N

CAS RN 6633-61-0

IUPAC Name: methyl 2-amino-1,3-thiazole-5-carboxylate
Synonyms: methyl 2-amino-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 158.18
Molecular Formula: C5H6N2O2S
InChi Key: UJNNCGWBDJHCEM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=C(N)S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results