accessibility menu, dialog, popup

UserName

Viewing profile as user:
(S)(R)(R)O(R)NNOFNH2OHHOMe

CAS RN 66335-38-4

IUPAC Name: 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-1,2-dihydropyrimidin-2-one
Synonyms: 5'-deoxy-5-fluorocytidine
Molecular Weight (g/mol): 245.21
Molecular Formula: C9H12FN3O4
InChi Key: YSNABXSEHNLERR-ZIYNGMLESA-N
SMILES: C[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=C(F)C(N)=NC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)