accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNNNH2

CAS RN 66346-91-6

IUPAC Name: 6-(morpholin-4-yl)pyridazin-3-amine
Synonyms: 6-(morpholin-4-yl)pyridazin-3-amine
Molecular Weight (g/mol): 180.21
Molecular Formula: C8H12N4O
InChi Key: IEUHTZVAUDMKQJ-UHFFFAOYSA-N
SMILES: NC1=NN=C(C=C1)N1CCOCC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)