accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONHONOO

CAS RN 6635-31-0

IUPAC Name: 5-nitro-6-oxo-1,6-dihydropyridine-3-carboxylic acid
Synonyms: 5-nitro-6-oxo-1H-pyridine-3-carboxylic acid
Molecular Weight (g/mol): 184.11
Molecular Formula: C6H4N2O5
InChi Key: VJMXJGVEVASJOD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CNC(=O)C(=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results